4-Chlorophenethyl bromide 95%
ACRO440180050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥94.0 %
: 1-(2-Bromoethyl)-4-chlorobenzene
:
:219.51 g/mol
:85 °C (1Torr)
:1.508 g/cm³
:>110 °C
:3082
:9,III
:GHS07
:GHS09
:Ambient
:6529-53-9
:C₈H₈BrCl
:MFCD00634271