

2-Fluorophenethyl bromide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:91319-54-9
:
Formula:C₈H₈BrF
MDL Number:MFCD04117466
:203.05 g/mol
Boiling Pt:73…75 °C (20Torr)
UN:3082
ADR:9,III
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together