Order Entry
Czech Republic
ContactUsLinkComponent
Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥98%
Irritant
Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥98%
Certificates
Irritant
Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥98%

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    100°C to 106°C
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together