


Pentafluorophenylmagnesium bromide 0.5 M in diethyl ether, AcroSeal®
Catalog # 438741000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Grey to brown to black Liquid
- Titration Grignard:0.4 to 0.6 M
Specification Test Results
About this item
:
:271.267 g/mol
Density:0.86 g/cm³
Flash Pt:–40 °C
Storage Temperature:Ambient
UN:2924
ADR:3,I
:GHS02
:GHS05
:GHS07
CAS Number:879-05-0
Formula:C₆BrF₅Mg
MDL Number:MFCD00015720