

Strontium nitrate 98%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10042-76-9
:
Formula:Sr(NO₃)₂
MDL Number:MFCD00011248
:211.63 g/mol
Boiling Pt:645 °C (1013 hPa)
Melting Pt:570 °C
Density:2,113 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1507
ADR:5.1,III
:GHS05
:GHS03
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Titration Complexometric:≥97.5 %
- Loss on drying:≤0.5 %
- pH:5 to 7 (5 % at 20°C)
- Heavy metals (as Pb):≤10 ppm
Specification Test Results
Frequently Bought Together