
2-Methylpropane-1-sulphonyl chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35432-36-1
:
Formula:C₄H₉ClO₂S
MDL Number:MFCD07369800
:156.624 g/mol
Boiling Pt:77…78 °C (11Torr)
Flash Pt:88 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear colorless to brown Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together