Order Entry
Czech Republic
ContactUsLinkComponent
1,4-Dioxane-2,3-diol 98%
Irritant
1,4-Dioxane-2,3-diol 98%
Catalog # ACRO438370050
CAS Number:  
undefined
Irritant
1,4-Dioxane-2,3-diol 98%
Catalog # ACRO438370050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Elemental Analysis:
    39.10 to 41.10 % (C)

Specification Test Results

About this item

 :
 :120.10 g/mol
Melting Pt:92…94 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:4845-50-5
Formula:C₄H₈O₄
MDL Number:MFCD00006572