
tert-Butyl-3-(trifluoromethylsulphonyloxy)-8-azabicyclo[3.2.1]oct-3-ene-8-carboxylate 95%
Catalog # ACRO438590010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder or low melting solid
- Infrared spectrum:Conforms
- GC:≥94.0 %
Specification Test Results
About this item
:
:357.344 g/mol
Melting Pt:36…40 °C
:GHS07
Storage Temperature:Ambient
CAS Number:185099-68-7
Formula:C₁₃H₁₈F₃NO₅S
MDL Number:MFCD14635483