
1-Boc-4-(2-chloroethyl)piperazine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:208167-83-3
:
Formula:C₁₁H₂₁ClN₂O₂
MDL Number:MFCD08443993
:248.751 g/mol
Melting Pt:62.9…64.7 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:62 to 68 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together