
O-(Tetrahydropyran-2-yl)-hydroxylamine 96%
ACRO437850010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White Low melting solid
- Infrared spectrum:Conforms
- Melting point:32 to 38 °C
- Water:≤1.0 %
- GC:≥95.0 %
:
:117.148 g/mol
:32.7…37 °C
:GHS07
:Ambient
:6723-30-4
:C₅H₁₁NO₂
:MFCD01321374