
(S)-3-(Aminomethyl)-1-boc-piperidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:140645-24-5
:
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD03839877
:214.31 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Specific optical rotation:-10° to -20° (20°C, 589 nm) (c=1, methanol)
Specification Test Results
Frequently Bought Together