Order Entry
Czech Republic
ContactUsLinkComponent
5-Bromo-1H-pyrrolo[2,3-b]pyridine 97%
Corrosive
Irritant
5-Bromo-1H-pyrrolo[2,3-b]pyridine 97%
Certificates
undefined
Corrosive
Irritant
5-Bromo-1H-pyrrolo[2,3-b]pyridine 97%

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    177 to 182 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together