Order Entry
Czech Republic
ContactUsLinkComponent
Chromium(III) nitrate nonahydrate 99.99% (trace metals basis)
Irritant
Oxidizing
Environment
Chromium(III) nitrate nonahydrate 99.99% (trace metals basis)
Catalog # ACRO437100050
CAS Number:  
undefined
Irritant
Oxidizing
Environment
Chromium(III) nitrate nonahydrate 99.99% (trace metals basis)
Catalog # ACRO437100050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Violet to black Crystals
  • Titration Iodometric:
    12.7 to 13.2 % (Cr)
  • Total trace metal impurities:
    ≤100 ppm

Specification Test Results

About this item

 :
 :400.15 g/mol
Melting Pt:60 °C
Density:1,807 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2720
ADR:5.1,III
 :GHS07
 :GHS03
 :GHS09
CAS Number:7789-02-8
Formula:CrN₃O₉·9H₂O
MDL Number:MFCD00149659