Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium hexachlororuthenate(IV) 99.99% (trace metals basis)
Toxic
Ammonium hexachlororuthenate(IV) 99.99% (trace metals basis)
Catalog # ACRO436930010
CAS Number:  
undefined
Toxic
Ammonium hexachlororuthenate(IV) 99.99% (trace metals basis)
Catalog # ACRO436930010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Dark brown to black Powder
  • Total trace metal impurities:
    ≤100 ppm

Specification Test Results

About this item

 :
 :349.86 g/mol
Storage Temperature:Ambient
UN:3288
ADR:6.1,III
 :GHS06
CAS Number:18746-63-9
Formula:H₈Cl₆N₂Ru
MDL Number:MFCD00015962