
2-Cyanopyrazine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19847-12-2
:
Formula:C₅H₃N₃
MDL Number:MFCD00049361
:105.10 g/mol
Boiling Pt:90…93 °C (15 mmHg)
Melting Pt:20 °C
Density:1.174 g/cm³
Flash Pt:97 °C
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5330 to 1.5350 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together