
Tetrabenzyl pyrophosphate 98%
Catalog # ACRO436750010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:59 to 65 °C
- Water:≤0.1 %
- HPLC:≥97.5 %
Specification Test Results
About this item
CAS Number:990-91-0
:
Formula:C₂₈H₂₈O₇P₂
MDL Number:MFCD00051941
:538.48 g/mol
Melting Pt:60.5…63 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05