Order Entry
Czech Republic
ContactUsLinkComponent
Tetrabutylammonium dihydrogen phosphate 0.4 M in acetonitrile, AcroSeal®
Flammable
Irritant
Tetrabutylammonium dihydrogen phosphate 0.4 M in acetonitrile, AcroSeal®
Catalog # 435991000.
CAS Number:  
undefined
Flammable
Irritant
Tetrabutylammonium dihydrogen phosphate 0.4 M in acetonitrile, AcroSeal®
Catalog # 435991000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to light yellow Solution
  • Titration with NaOH:
    0.38 to 0.45 M
  • Melting point:
    215 to 220 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :339.46 g/mol
Boiling Pt:81…82 °C (760 mmHg)
Melting Pt:–48 °C
Density:0.828 g/cm³
Flash Pt:2 °C
UN:1648
ADR:3,II
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:5574-97-0
Formula:C₁₆H₃₈NO₄P
MDL Number:MFCD00064526