Order Entry
Czech Republic
ContactUsLinkComponent
Tetraisobutyldialuminoxane 10% (w/w) in toluene, AcroSeal®
Flammable
Irritant
Health
Tetraisobutyldialuminoxane 10% (w/w) in toluene, AcroSeal®
Catalog # 435988000.
CAS Number:  
undefined
Flammable
Irritant
Health
Tetraisobutyldialuminoxane 10% (w/w) in toluene, AcroSeal®
Catalog # 435988000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    800 mL
  • Seq:
    2805

Specifications

Specification Test Results

  • Appearance:
    Colorless Solution
  • Titration Complexometric:
    9.5 to 11.0 % w/w
  • Refractive index:
    1.5550 to 1.5590 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
Density:0.867 g/cm³
Flash Pt:–8 °C
ADR:3,II
 :298.426 g/mol
UN:1993
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:998-00-5
Formula:C₁₆H₃₆Al₂O
MDL Number:MFCD00192480