
Tetrabutylammonium hydroxide-30-hydrate 95+%
:147741-30-8
:
:C₁₆H₉₇NO₃₁
:MFCD00149573
:799.928 g/mol
:27…30 °C
:Refrigerator
:3263
:8,II
:GHS05
- Appearance:Transparent white Crystals or crystalline powder
- Melting point:26 to 30°C
- Titration with HCl:≥95.0 %
- HPLC:≥96.0 %
Frequently Bought Together