Order Entry
Czech Republic
ContactUsLinkComponent
(R)-tert-Butyl(1,4-dihydroxybutan-2-yl)carbamate 97%
 
(R)-tert-Butyl(1,4-dihydroxybutan-2-yl)carbamate 97%
  ACRO435570050
 :  
undefined
 
(R)-tert-Butyl(1,4-dihydroxybutan-2-yl)carbamate 97%
  ACRO435570050
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    65 to 69 °C
  • GC:
    ≥96.0 %
  • Enantiomeric excess:
    ≥98.0 %
  • Specific optical rotation:
    +7° to +9° (20 °C, 589 nm) (c=1, chloroform)

 

 

 :
 :205.25 g/mol
 :66.4…72.1 °C
 :GHS07
 :Ambient
 :397246-14-9
 :C₉H₁₉NO₄
 :MFCD08703679