Order Entry
Czech Republic
ContactUsLinkComponent
1-Propenylmagnesium bromide 0.5 M in THF, AcroSeal®
1-Propenylmagnesium bromide 0.5 M in THF, AcroSeal®
Certificates
undefined
1-Propenylmagnesium bromide 0.5 M in THF, AcroSeal®

Specification Test Results

  • Appearance:
    Light yellow to orange to brown Solution
  • Titration Grignard:
    0.45 to 0.55 M
  • Melting point:
    138 to 142 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Frequently Bought Together