
n-Dodecane, anhydrous 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-40-3
:
Formula:H₃C(CH₂)₁₀CH₃
MDL Number:MFCD00008969
:170.34 g/mol
Boiling Pt:215…217 °C (760 mmHg)
Melting Pt:–12 °C
Density:0.753 g/cm³
Flash Pt:71 °C
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥98.5 %
Specification Test Results
Related Information
Frequently Bought Together