


Dilithium tetrabromonickelate(II) 0.5 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13826-95-4
:
Formula:Br₄Li₂Ni
MDL Number:MFCD00192058
Density:1.065 g/cm³
Flash Pt:–18 °C
Storage Temperature:Ambient
UN:1993
ADR:3,II
:392.189 g/mol
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Blue-green to brown Solution
- Titration Argentometric:0.45 to 0.55 M
- GC:≥95 %
Specification Test Results
Related Information
Frequently Bought Together