
(R)-(+)-3-Hydroxypiperidine hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:198976-43-1
:
Formula:C₅H₁₂ClNO
MDL Number:MFCD00192221
:137.607 g/mol
Melting Pt:134…138 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Additional info:May contain up to 3% water
- Elemental Analysis:42.18 to 44.73 % C
- Specific optical rotation:+5° to +8° (20°C, 589 nm) (c=2, methanol)
Specification Test Results
Frequently Bought Together