(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane 97%
Catalog # ACRO434121000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- NMR:≥96.0 %
- GC:≥99.5 %
- Residue after evaporation:≤0.001 %
Specification Test Results
About this item
:
:292.36 g/mol
Flash Pt:>110 °C
Storage Temperature:Ambient
CAS Number:1314246-02-0
Formula:C₁₇H₂₅O₂P
MDL Number:MFCD09265099