


Piperidine 99.5%, AcroSeal®, purified by redistillation
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-89-4
:
Formula:C₅H₁₁N
MDL Number:MFCD00005979
:85.15 g/mol
Boiling Pt:107.3 °C (1013 hPa)
Melting Pt:–11 °C
Density:0.862 g/cm³
Flash Pt:16 °C
UN:2401
ADR:8,I
:GHS06
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4510 to 1.4540 (20°C, 589 nm)
- GC:≥99.4 %
- Color scale:≤20 APHA
Specification Test Results
Related Information
Frequently Bought Together