



Cyclohexylmethylmagnesium bromide 0.25 M in THF, AcroSeal®
Catalog # 433611000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless to yellow to brown Solution
- Titration Grignard:0.22 to 0.3 M
- Water:≤0.005 %
- GC:≥99.0 %
- Residue after evaporation:≤0.001 %
- Peroxides:≤20 ppm
Specification Test Results
About this item
:
:201.39 g/mol
Density:0.966 g/cm³
Flash Pt:–20 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:35166-78-0
Formula:C₇H₁₃BrMg
MDL Number:MFCD02260210