
(3,5-Dichlorophenyl)methanesulphonyl chloride 97%
ACRO433230010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to light yellow Crystalline powder or solid
- Infrared spectrum:Conforms
- Melting point:106 to 112 °C
- HPLC:≥96.0 % as diethylsulfonamide derivative
:
:259.54 g/mol
:108.6…110.6 °C
:Ambient
:3261
:8,II
:GHS05
:163295-70-3
:C₇H₅CI₃O₂S
:MFCD01631928