Order Entry
Czech Republic
ContactUsLinkComponent
1-Hexanol, anhydrous 99%, AcroSeal®
Flammable
Irritant
1-Hexanol, anhydrous 99%, AcroSeal®
Certificates
undefined
Flammable
Irritant
1-Hexanol, anhydrous 99%, AcroSeal®

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.005 %
  • GC:
    ≥98.5 %
  • Residue after evaporation:
    ≤0.002 %

Specification Test Results

Frequently Bought Together