


Heptylmagnesium bromide 1 M in diethyl ether, AcroSeal®
Catalog # 433031000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Brown Solution
- Titration Grignard:0.95 to 1.10 M
- Titration Argentometric:96.0 to 104.0 %
Specification Test Results
About this item
:
:203.41 g/mol
Boiling Pt:35 °C (760 mmHg)
Density:0.841 g/cm³
Flash Pt:–30 °C
Storage Temperature:Ambient
UN:2924
ADR:3,I
:GHS02
:GHS05
:GHS07
CAS Number:13125-66-1
Formula:C₇H₁₅BrMg
MDL Number:MFCD00013517