
2-Methylsulphanylpyrimidine-4-carboxylic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1126-44-9
:
Formula:C₆H₆N₂O₂S
MDL Number:MFCD09834808
:170.19 g/mol
Melting Pt:208…209 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together