Order Entry
Czech Republic
ContactUsLinkComponent
n-Tetratriacontane 98%
n-Tetratriacontane 98%
Catalog # ACRO432070010
CAS Number:  
undefined
n-Tetratriacontane 98%
Catalog # ACRO432070010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White Flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    72 to 76 °C
  • Elemental Analysis:
    83.14 to 87.40 % C

Specification Test Results

About this item

 :
 :478.93 g/mol
Boiling Pt:482 °C (1013 hPa)
Melting Pt:73.9…74.9 °C
Density:0,7728 g/cm³ (90 °C)
Storage Temperature:Ambient
CAS Number:14167-59-0
Formula:H₃C(CH₂)₃₂CH₃
MDL Number:MFCD00009412