Order Entry
Czech Republic
ContactUsLinkComponent
(R)-(+)-3-Chloro-1-phenyl-1-propanol 97%
Irritant
(R)-(+)-3-Chloro-1-phenyl-1-propanol 97%
Certificates
undefined
Irritant
(R)-(+)-3-Chloro-1-phenyl-1-propanol 97%

Specification Test Results

  • Appearance:
    White to yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    58 to 61 °C
  • GC:
    ≥96.0 %
  • Specific optical rotation:
    +23° to +29° (20 °C, 589 nm) (c=1 in Chloroform)

Specification Test Results

Related Information

Frequently Bought Together