



5-Fluoro-2-methylphenylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 431830500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Typ balení:Glass bottle, AcroSeal®
- Bal.:50 mL
- Sekvence:2370
Specifications
Specification Test Results
- Appearance:Clear yellow Solution
- Titration Grignard:0.40 to 0.60 M
- Titration with HClO4:≥98.0 %
Specification Test Results
About this item
:
:213.332 g/mol
Density:0,96 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:186496-59-3
Formula:C₇H₆BrFMg
MDL Number:MFCD01311468