

Dicyclohexylmethane-4,4'-diisocyanate (mixture of isomers) mixture of isomers 90% (mixture of isomers), AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5124-30-1
:
Formula:C₁₅H₂₂N₂O₂
MDL Number:MFCD00015405
:262.35 g/mol
Boiling Pt:112…114 °C (1010hPa)
Melting Pt:19…23 °C
Density:1.066 g/cm³
Flash Pt:113 °C
Storage Temperature:Ambient
UN:2206
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4960 to 1.4980 (20°C, 589 nm)
- GC:≥88 %
- Stabilizer:0.05 % BHT
Specification Test Results
Related Information
Frequently Bought Together