



9-Phenanthrylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 431730500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Clear yellow to brown Solution
- Titration Grignard:0.40 to 0.60 M
- Residue after ign. (sulfated):≤0.01 %
- Phosphate (PO4):≤0.002 %
- Sulfate (SO4):≤0.002 %
- Calcium (Ca):≤0.001 %
- Iron (Fe):≤0.0005 %
- Magnesium (Mg):≤0.001 %
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.01 % (5 % in water)
Specification Test Results
About this item
:
:281.435 g/mol
Density:0.945 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:71112-64-6
Formula:C₁₄H₉BrMg
MDL Number:MFCD00672008