



[4-(1-Piperidinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
Catalog # 431690500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Brown Solution
- Titration Grignard:0.15 to 0.35 M
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:278.476 g/mol
Density:0.924 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:480424-83-7
Formula:C₁₂H₁₆BrMgN
MDL Number:MFCD03840896