Order Entry
Czech Republic
ContactUsLinkComponent
1,3-Dioxolane, anhydrous 99.8% stabilised, AcroSeal®
Flammable
Irritant
1,3-Dioxolane, anhydrous 99.8% stabilised, AcroSeal®
Certificates
undefined
Flammable
Irritant
1,3-Dioxolane, anhydrous 99.8% stabilised, AcroSeal®

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 %
  • GC:
    ≥99.75 %
  • Stabilizer:
    75 ppm BHT
  • Residue after ignition:
    ≤0.0005 %

Specification Test Results

Frequently Bought Together