Order Entry
Czech Republic
ContactUsLinkComponent
cis-1,4-Diacetoxy-2-butene 95%
Irritant
cis-1,4-Diacetoxy-2-butene 95%
Catalog # ACRO431260250
CAS Number:  
undefined
Irritant
cis-1,4-Diacetoxy-2-butene 95%
Catalog # ACRO431260250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥94.0 %

Specification Test Results

About this item

 :
 :172.18 g/mol
Boiling Pt:222…223 °C
Density:1.08 g/cm³
Flash Pt:>113 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:25260-60-0
Formula:C₈H₁₂O₄
MDL Number:MFCD00059339