


3-Bromobenzylmagnesium bromide 0.25 M in diethyl ether, AcroSeal®
Catalog # 431601000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear yellow Solution
- Titration Grignard:0.15 to 0.35 M
- Titration with HClO4:≥90.0 %
Specification Test Results
About this item
:
:274.238 g/mol
Density:0.766 g/cm³
Flash Pt:–40 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS02
CAS Number:107549-22-4
Formula:C₇H₆Br₂Mg
MDL Number:MFCD01319902