

2-(4-Fluorophenyl)propan-2-amine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17797-10-3
:
Formula:C₉H₁₂FN
MDL Number:MFCD07777176
:153.2 g/mol
Boiling Pt:40 °C (7mbar)
Flash Pt:92 °C
Storage Temperature:Refrigerator
UN:2735
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together