



(±)-Epichlorhydrin 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-89-8
:
Formula:C₃H₅ClO
MDL Number:MFCD00005132
:92.52 g/mol
Boiling Pt:115…117 °C (760 mmHg)
Melting Pt:–57 °C
Density:1.18 g/cm³
Flash Pt:28 °C
UN:2023
ADR:6.1,II
:GHS06
:GHS08
:GHS05
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4365 to 1.4385 (20°C, 589 nm)
- Water:≤0.02 % (K.F.)
- GC:≥98.5 %
- Color scale:≤15 APHA
Specification Test Results
Related Information
Certificates
Search by Lot NumberACRO43071
SDS
AcroSeal Brochure VWR Final AO_AcroSeal_Product Lists_VWR_final
Product Literature
Frequently Bought Together