
3-Methoxy-5-(trifluoromethyl)phenylboronic acid 97%
:871332-97-7
:
:C₈H₈BF₃O₃
:MFCD08056362
:219.95 g/mol
:158.4…160.8 °C
:GHS07
:Refrigerator
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:157 to 162 °C
- HPLC:≥96.0 %
Frequently Bought Together