
1H-Indazole-7-carboxylic acid 97%
ACRO429600010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:228 to 233 °C
- HPLC:≥96.0 %
:
:162.15 g/mol
:229…231 °C
:GHS07
:Ambient
:677304-69-7
:C₈H₆N₂O₂
:MFCD06804572