
2-Bromothiazole-4-carbaldehyde 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5198-80-1
:
Formula:C₄H₂BrNOS
MDL Number:MFCD04115726
:192.03 g/mol
Melting Pt:128…130 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:126 to 130 °C
- HPLC:≥96.0 %
- UV:1 cm cell vs HPLC-grade water
- Non-volatile matter:≤0.0003 %
- Acidity:≤0.0002 meq/g
- Alkalinity:≤0.0002 meq/g
Specification Test Results
Frequently Bought Together