Order Entry
Czech Republic
ContactUsLinkComponent
Propylene carbonate, anhydrous 99.5%, AcroSeal®
Propylene carbonate, anhydrous 99.5%, AcroSeal®
Certificates
undefined
Propylene carbonate, anhydrous 99.5%, AcroSeal®

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 %
  • GC:
    ≥99.4 %

Specification Test Results

Frequently Bought Together