

Butyl acetate, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-86-4
:
Formula:CH₃COO(CH₂)₃CH₃
MDL Number:MFCD00009445
:116.16 g/mol
Boiling Pt:127 °C (760 mmHg)
Melting Pt:–78 °C
Density:0.88 g/cm³
Flash Pt:27 °C
Storage Temperature:Ambient
UN:1123
ADR:3,III
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
Specification Test Results
Related Information
Frequently Bought Together