Order Entry
Czech Republic
ContactUsLinkComponent
 
Toxic
Health
Environment
Flammable
(Trimethylsilyl)diazomethane ∼2.0 M in hexanes, AcroSeal®, Technical Grade
Catalog # 429211000.
CAS Number:  
undefined
Toxic
Health
Environment
Flammable
(Trimethylsilyl)diazomethane ∼2.0 M in hexanes, AcroSeal®, Technical Grade
Catalog # 429211000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear yellow Solution
  • Strength:
    1.90 to 2.40 M
  • Sulfate (SO4):
    ≤30 ppm
  • Iron (Fe):
    ≤10 ppm

Specification Test Results

About this item

 :
Boiling Pt:96 °C (760 mmHg)
Density:0.675 g/cm³
Flash Pt:–23 °C
ADR:3,II
 :114.22 g/mol
UN:1992
 :GHS06
 :GHS08
 :GHS09
 :GHS02
Storage Temperature:Ambient
CAS Number:18107-18-1
Formula:C₄H₁₀N₂Si
MDL Number:MFCD00053946