


Tetrahydroboritan sodný 12% in sodium hydroxide solution 40%, AcroSeal®
Catalog # 429131000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Typ balení:Glass bottle, AcroSeal®
- Bal.:100 mL
- Sekvence:2440
Specifications
Specification Test Results
- Appearance:Colorless Liquid
- Titration Iodometric:11 to 13 %
- GC:≥95.0 %
Specification Test Results
About this item
CAS Number:16940-66-2
:
Formula:NaBH₄
MDL Number:MFCD00003518
Boiling Pt:130…135 °C (760 mmHg)
Melting Pt:400 °C
Density:1,4 g/cm³
Flash Pt:69 °C
UN:3320
ADR:8,II
:GHS07
:GHS05
:GHS08
Storage Temperature:Ambient