Order Entry
Czech Republic
ContactUsLinkComponent
Propanephosphonic acid cyclic anhydride 50% (w/w) in DMF, anhydrous, AcroSeal®
Flammable
Corrosive
Irritant
Health
Propanephosphonic acid cyclic anhydride 50% (w/w) in DMF, anhydrous, AcroSeal®
Catalog # 429111000.
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Propanephosphonic acid cyclic anhydride 50% (w/w) in DMF, anhydrous, AcroSeal®
Catalog # 429111000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to brown Solution
  • Refractive index:
    1.4500 to 1.4540 (20°C, 589 nm)
  • Assay:
    ≥50 % Solids

Specification Test Results

About this item

 :
 :318.19 g/mol
Boiling Pt:153 °C
Density:1.09 g/cm³
Flash Pt:56 °C
Storage Temperature:Ambient
UN:2920
ADR:8,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:68957-94-8
Formula:C₉H₂₁O₆P₃
MDL Number:MFCD00006583